Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFYVE21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198953.1 | NP_001185882.1 | 6 - 7 | 684 | 115 | |
GenBank mRNA | AK057816.1 | - | 6 - 7 | 605 | 115 | |
AK226142.1 | - | 4 - 5 | 3647 | 115 | ||
CX786145.1 | - | 6 - 7 | 684 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |