Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001624.3 | NP_001615.2 | 13 - 14 | 4737 | 68 | |
XM_005266839.3 | XP_005266896.1 | 15 - 16 | 5575 | 68 | ||
XM_017010333.1 | XP_016865822.1 | 13 - 14 | 4962 | 68 | ||
GenBank mRNA | AK025693.1 | - | 2 - 3 | 111 | 68 | |
AK297008.1 | - | 3 - 4 | 297 | 68 | ||
AK300443.1 | - | 7 - 8 | 772 | 68 | ||
AK316287.1 | - | 7 - 8 | 773 | 68 | ||
BC062788.1 | - | 7 - 8 | 795 | 68 | ||
CV573838.1 | - | 2 - 3 | 149 | 68 | ||
U83115.1 | - | 13 - 14 | 3992 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons |