Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KMT2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006718839.3 | XP_006718902.2 | 2 - 3 | 523 | 71 | |
XM_011542833.2 | XP_011541135.1 | 3 - 4 | 622 | 71 | ||
GenBank mRNA | AY187943.1 | - | 187 | 71 | ||
AY187945.1 | - | 316 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Homo sapiens Probe spans exons |