Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUC16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024690.2 | NP_078966.2 | 13 - 14 | 36869 | 71 | |
XM_017027486.1 | XP_016882975.1 | 17 - 18 | 36885 | 71 | ||
XM_017027487.1 | XP_016882976.1 | 17 - 18 | 36885 | 71 | ||
XM_017027488.1 | XP_016882977.1 | 17 - 18 | 36885 | 71 | ||
XM_017027489.1 | XP_016882978.1 | 14 - 15 | 36731 | 71 | ||
XM_017027490.1 | XP_016882979.1 | 17 - 18 | 36885 | 71 | ||
XM_017027491.1 | XP_016882980.1 | 17 - 18 | 36885 | 71 | ||
XM_017027492.1 | XP_016882981.1 | 17 - 18 | 36885 | 71 | ||
XM_017027493.1 | XP_016882982.1 | 17 - 18 | 36885 | 71 | ||
XM_017027494.1 | XP_016882983.1 | 17 - 18 | 36885 | 71 | ||
XM_017027495.1 | XP_016882984.1 | 17 - 18 | 36885 | 71 | ||
XM_017027496.1 | XP_016882985.1 | 17 - 18 | 36885 | 71 | ||
XM_017027497.1 | XP_016882986.1 | 17 - 18 | 36885 | 71 | ||
XM_017027498.1 | XP_016882987.1 | 17 - 18 | 36885 | 71 | ||
XM_017027499.1 | XP_016882988.1 | 17 - 18 | 27672 | 71 | ||
XM_017027500.1 | XP_016882989.1 | 16 - 17 | 27591 | 71 | ||
GenBank mRNA | AF361486.3 | - | 11783 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Probe spans exons |