Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010898.hg.1 |
ENST00000327438.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010898.hg.1 |
ENST00000330163.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010898.hg.1 |
ENST00000338354.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010898.hg.1 |
ENST00000339712.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010898.hg.1 |
ENST00000339871.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010898.hg.1 |
ENST00000341278.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010898.hg.1 |
ENST00000344338.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010898.hg.1 |
ENST00000344467.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010898.hg.1 |
ENST00000359586.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010898.hg.1 |
ENST00000368909.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010898.hg.1 |
ENST00000368915.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010898.hg.1 |
ENST00000368942.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010898.hg.1 |
ENST00000368953.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010898.hg.1 |
ENST00000368955.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010898.hg.1 |
ENST00000368956.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010898.hg.1 |
NM_004406.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010898.hg.1 |
NM_007329.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010898.hg.1 |
NM_017579.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010898.hg.1 |
TC10000890.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000162781.hg.1 |
ENST00000330163.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162781.hg.1 |
ENST00000338354.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162781.hg.1 |
ENST00000344338.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162781.hg.1 |
ENST00000359586.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162781.hg.1 |
ENST00000368909.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162781.hg.1 |
ENST00000368955.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162781.hg.1 |
ENST00000368956.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162781.hg.1 |
ENST00000619379.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162781.hg.1 |
NM_004406.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000162781.hg.1 |
NM_007329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000162781.hg.1 |
NM_017579.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000162781.hg.1 |
ENST00000330163.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162781.hg.1 |
ENST00000338354.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162781.hg.1 |
ENST00000344338.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162781.hg.1 |
ENST00000359586.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162781.hg.1 |
ENST00000368909.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162781.hg.1 |
ENST00000368955.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162781.hg.1 |
ENST00000368956.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162781.hg.1 |
ENST00000619379.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000162781.hg.1 |
NM_004406.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000162781.hg.1 |
NM_007329.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000162781.hg.1 |
NM_017579.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000162781.hg.1 |
TC1000009155.hg.1 |
Clariom D TC |