Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001624.3 | NP_001615.2 | 3 - 4 | 3556 | 131 | |
XM_005266839.3 | XP_005266896.1 | 5 - 6 | 4394 | 131 | ||
XM_017010333.1 | XP_016865822.1 | 3 - 4 | 3781 | 131 | ||
GenBank mRNA | U83115.1 | - | 3 - 4 | 2811 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons |