Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007249.hg.1 |
ENST00000381647.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007249.hg.1 |
ENST00000381647.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007249.hg.1 |
ENST00000381647.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007249.hg.1 |
ENST00000381647.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007249.hg.1 |
ENST00000381647.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007249.hg.1 |
ENST00000381647.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007249.hg.1 |
ENST00000381647.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007249.hg.1 |
ENST00000381647.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007249.hg.1 |
ENST00000381647.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007249.hg.1 |
ENST00000381647.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007249.hg.1 |
ENST00000505931.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007249.hg.1 |
ENST00000505931.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007249.hg.1 |
ENST00000505931.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007249.hg.1 |
ENST00000505931.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007249.hg.1 |
ENST00000505931.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007249.hg.1 |
ENST00000505931.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007249.hg.1 |
ENST00000509976.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007249.hg.1 |
ENST00000509976.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007249.hg.1 |
ENST00000509976.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007249.hg.1 |
ENST00000509976.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007249.hg.1 |
ENST00000509976.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007249.hg.1 |
ENST00000509976.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007249.hg.1 |
ENST00000612065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007249.hg.1 |
ENST00000612065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007249.hg.1 |
ENST00000612065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007249.hg.1 |
ENST00000612065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007249.hg.1 |
ENST00000612065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007249.hg.1 |
NM_175921.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007249.hg.1 |
NM_175921.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007249.hg.1 |
NM_175921.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007249.hg.1 |
NM_175921.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007249.hg.1 |
NM_175921.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007249.hg.1 |
NM_175921.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007249.hg.1 |
NM_175921.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007249.hg.1 |
NM_175921.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007249.hg.1 |
NM_175921.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007249.hg.1 |
NM_175921.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05002216.hg.1 |
TC05000189.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05002216.hg.1 |
ENST00000381647.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05002216.hg.1 |
ENST00000505931.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05002216.hg.1 |
NM_175921.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007249.hg.1 |
ENST00000381647.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007249.hg.1 |
ENST00000381647.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007249.hg.1 |
ENST00000381647.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007249.hg.1 |
ENST00000381647.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007249.hg.1 |
ENST00000381647.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007249.hg.1 |
ENST00000381647.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007249.hg.1 |
ENST00000381647.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007249.hg.1 |
ENST00000381647.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007249.hg.1 |
ENST00000381647.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007249.hg.1 |
ENST00000381647.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007249.hg.1 |
ENST00000505931.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007249.hg.1 |
ENST00000505931.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007249.hg.1 |
ENST00000505931.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007249.hg.1 |
ENST00000505931.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007249.hg.1 |
ENST00000505931.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007249.hg.1 |
ENST00000505931.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007249.hg.1 |
ENST00000509976.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007249.hg.1 |
ENST00000509976.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007249.hg.1 |
ENST00000509976.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007249.hg.1 |
ENST00000509976.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007249.hg.1 |
ENST00000509976.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007249.hg.1 |
ENST00000509976.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007249.hg.1 |
ENST00000612065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007249.hg.1 |
ENST00000612065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007249.hg.1 |
ENST00000612065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007249.hg.1 |
ENST00000612065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007249.hg.1 |
ENST00000612065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007249.hg.1 |
NM_175921.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007249.hg.1 |
NM_175921.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007249.hg.1 |
NM_175921.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007249.hg.1 |
NM_175921.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007249.hg.1 |
NM_175921.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007249.hg.1 |
NM_175921.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007249.hg.1 |
NM_175921.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007249.hg.1 |
NM_175921.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007249.hg.1 |
NM_175921.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007249.hg.1 |
NM_175921.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148978.hg.1 |
ENST00000381647.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148978.hg.1 |
ENST00000505931.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148978.hg.1 |
ENST00000509976.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148978.hg.1 |
ENST00000612065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148978.hg.1 |
NM_175921.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148978.hg.1 |
ENST00000381647.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148978.hg.1 |
ENST00000505931.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148978.hg.1 |
ENST00000509976.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148978.hg.1 |
ENST00000612065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148978.hg.1 |
NM_175921.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148978.hg.1 |
TC0500007249.hg.1 |
Clariom D TC |