Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000449873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000449873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000449873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000449873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
ENST00000449873.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000207157.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000369429.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000449873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000449873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000449873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000449873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
ENST00000449873.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015445.hg.1 |
NM_152380.2 |
Refseq |