Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010103.hg.1 |
ENST00000298746.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010103.hg.1 |
ENST00000485065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010103.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010103.hg.1 |
TC10000837.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
ENST00000485065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
ENST00000485065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
ENST00000485065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
ENST00000485065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
ENST00000485065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010102.hg.1 |
ENST00000298746.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010102.hg.1 |
ENST00000485065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010102.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010102.hg.1 |
TC10000837.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010100.hg.1 |
ENST00000298746.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010100.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010100.hg.1 |
TC10000837.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010101.hg.1 |
ENST00000298746.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010101.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010101.hg.1 |
TC10000837.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000161471.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161471.hg.1 |
ENST00000485065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161471.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161471.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161471.hg.1 |
ENST00000485065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161471.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161471.hg.1 |
TC1000008988.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
ENST00000485065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
ENST00000485065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
ENST00000485065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
ENST00000485065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
ENST00000485065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161468.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161468.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161468.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161468.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161468.hg.1 |
TC1000008988.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000161469.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161469.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161469.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161469.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161469.hg.1 |
TC1000008988.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000161470.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161470.hg.1 |
ENST00000485065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161470.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161470.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161470.hg.1 |
ENST00000485065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161470.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161470.hg.1 |
TC1000008988.hg.1 |
Clariom D TC |