Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010108.hg.1 |
ENST00000298746.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010108.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010108.hg.1 |
TC10000837.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
ENST00000485065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
ENST00000485065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
ENST00000485065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
ENST00000485065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
ENST00000485065.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010109.hg.1 |
ENST00000298746.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010109.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10010109.hg.1 |
TC10000837.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000161476.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161476.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161476.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161476.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161476.hg.1 |
TC1000008988.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000161477.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161477.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161477.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000161477.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000161477.hg.1 |
TC1000008988.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
ENST00000298746.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
ENST00000485065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
ENST00000485065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
ENST00000485065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
ENST00000485065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
ENST00000485065.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000008988.hg.1 |
NM_139169.4 |
Refseq |