Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02003405.hg.1 |
TC02000230.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02003405.hg.1 |
ENST00000280527.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02003405.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000413985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000413985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000413985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000413985.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000426856.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000426856.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000426856.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000426856.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000426856.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000428774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000428774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000428774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000428774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000473403.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000473403.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000473403.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000477491.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000477491.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000477491.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000481321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000481321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000481321.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000497236.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
ENST00000497236.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200150096.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200150096.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200150096.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200150096.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200150096.hg.1 |
TC0200007237.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000280527.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000413985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000413985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000413985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000413985.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000426856.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000426856.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000426856.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000426856.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000426856.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000428774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000428774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000428774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000428774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000473403.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000473403.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000473403.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000477491.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000477491.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000477491.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000481321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000481321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000481321.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000497236.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
ENST00000497236.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007237.hg.1 |
NM_016441.2 |
Refseq |