Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X017383.hg.1 |
TC0X001284.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X017383.hg.1 |
ENST00000262839.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X017383.hg.1 |
NM_012471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X017382.hg.1 |
TC0X001284.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X017382.hg.1 |
ENST00000262839.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X017382.hg.1 |
NM_012471.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X017381.hg.1 |
TC0X001284.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X017381.hg.1 |
ENST00000262839.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X017381.hg.1 |
NM_012471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00171772.hg.1 |
ENST00000262839.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00171772.hg.1 |
NM_012471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00171772.hg.1 |
ENST00000262839.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00171772.hg.1 |
NM_012471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00171772.hg.1 |
TC0X00010525.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00171774.hg.1 |
ENST00000262839.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00171774.hg.1 |
NM_012471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00171774.hg.1 |
ENST00000262839.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00171774.hg.1 |
NM_012471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00171774.hg.1 |
TC0X00010525.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00171775.hg.1 |
ENST00000262839.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00171775.hg.1 |
NM_012471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00171775.hg.1 |
ENST00000262839.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00171775.hg.1 |
NM_012471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00171775.hg.1 |
TC0X00010525.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |