Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000527656.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000527656.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000527656.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000527656.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000529604.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000529604.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000529604.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000529604.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000529604.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000529921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000529921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000529921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000529921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000529921.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000534404.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000534404.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000534404.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000534404.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000534404.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
ENST00000534404.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11020132.hg.1 |
ENST00000378623.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11020132.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11020132.hg.1 |
TC11001728.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000527656.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000527656.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000527656.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000527656.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000529604.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000529604.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000529604.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000529604.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000529604.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000529921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000529921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000529921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000529921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000529921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000534404.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000534404.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000534404.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000534404.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000534404.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
ENST00000534404.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010718.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100181096.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100181096.hg.1 |
ENST00000529921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100181096.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100181096.hg.1 |
ENST00000378623.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100181096.hg.1 |
ENST00000529921.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100181096.hg.1 |
NM_002334.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100181096.hg.1 |
TC1100010718.hg.1 |
Clariom D TC |