Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
ENST00000263921.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
ENST00000263921.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
ENST00000263921.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
ENST00000263921.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
ENST00000263921.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
ENST00000263921.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
ENST00000263921.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
ENST00000263921.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
ENST00000263921.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
ENST00000263921.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
ENST00000263921.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
ENST00000263921.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
ENST00000263921.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
ENST00000509678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
ENST00000509678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
ENST00000509678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
ENST00000509678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
ENST00000509678.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
ENST00000510894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
ENST00000510894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
ENST00000510894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
ENST00000510894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
ENST00000510894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
ENST00000510894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
ENST00000510894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
ENST00000512964.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
ENST00000512964.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
ENST00000512964.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
ENST00000512964.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
ENST00000512964.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
ENST00000512964.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
ENST00000512964.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
ENST00000512964.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
NM_012110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
NM_012110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
NM_012110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
NM_012110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
NM_012110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
NM_012110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
NM_012110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
NM_012110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
NM_012110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
NM_012110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
NM_012110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010715.hg.1 |
NM_012110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016482.hg.1 |
TC04001201.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016482.hg.1 |
ENST00000263921.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016482.hg.1 |
ENST00000512964.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016482.hg.1 |
NM_012110.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016481.hg.1 |
TC04001201.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016481.hg.1 |
ENST00000263921.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016481.hg.1 |
ENST00000512964.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04016481.hg.1 |
NM_012110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172943.hg.1 |
ENST00000263921.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172943.hg.1 |
ENST00000509678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172943.hg.1 |
ENST00000512964.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172943.hg.1 |
NM_012110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172943.hg.1 |
ENST00000263921.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172943.hg.1 |
ENST00000509678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172943.hg.1 |
ENST00000512964.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172943.hg.1 |
NM_012110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172943.hg.1 |
TC0400010715.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
ENST00000263921.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
ENST00000263921.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
ENST00000263921.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
ENST00000263921.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
ENST00000263921.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
ENST00000263921.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
ENST00000263921.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
ENST00000263921.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
ENST00000263921.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
ENST00000263921.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
ENST00000263921.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
ENST00000263921.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
ENST00000263921.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
ENST00000509678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
ENST00000509678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
ENST00000509678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
ENST00000509678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
ENST00000509678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
ENST00000510894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
ENST00000510894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
ENST00000510894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
ENST00000510894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
ENST00000510894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
ENST00000510894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
ENST00000510894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
ENST00000512964.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
ENST00000512964.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
ENST00000512964.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
ENST00000512964.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
ENST00000512964.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
ENST00000512964.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
ENST00000512964.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
ENST00000512964.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
NM_012110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
NM_012110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
NM_012110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
NM_012110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
NM_012110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
NM_012110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
NM_012110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
NM_012110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
NM_012110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
NM_012110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
NM_012110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010715.hg.1 |
NM_012110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172944.hg.1 |
ENST00000263921.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172944.hg.1 |
ENST00000509678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172944.hg.1 |
ENST00000510894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172944.hg.1 |
ENST00000512964.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172944.hg.1 |
NM_012110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172944.hg.1 |
ENST00000263921.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172944.hg.1 |
ENST00000509678.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172944.hg.1 |
ENST00000510894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172944.hg.1 |
ENST00000512964.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400172944.hg.1 |
NM_012110.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400172944.hg.1 |
TC0400010715.hg.1 |
Clariom D TC |