Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14015545.hg.1 |
ENST00000261258.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14015545.hg.1 |
ENST00000336735.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14015545.hg.1 |
NM_005065.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14015545.hg.1 |
TC14001372.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14015547.hg.1 |
ENST00000261258.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14015547.hg.1 |
ENST00000336735.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14015547.hg.1 |
NM_005065.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR14015547.hg.1 |
TC14001372.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1400172656.hg.1 |
ENST00000336735.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400172656.hg.1 |
ENST00000554293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400172656.hg.1 |
NM_005065.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400172656.hg.1 |
ENST00000336735.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400172656.hg.1 |
ENST00000554293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400172656.hg.1 |
NM_005065.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400172656.hg.1 |
TC1400009842.hg.1 |
Clariom D TC |