Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004163.hg.1 |
TC14002198.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004163.hg.1 |
ENST00000216294.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR14004163.hg.1 |
NM_003082.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010620.hg.1 |
ENST00000216294.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010620.hg.1 |
ENST00000216294.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010620.hg.1 |
ENST00000216294.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010620.hg.1 |
ENST00000216294.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010620.hg.1 |
ENST00000216294.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010620.hg.1 |
ENST00000216294.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010620.hg.1 |
ENST00000216294.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010620.hg.1 |
ENST00000216294.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010620.hg.1 |
ENST00000216294.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010620.hg.1 |
ENST00000216294.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010620.hg.1 |
ENST00000216294.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010620.hg.1 |
ENST00000216294.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010620.hg.1 |
NM_003082.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010620.hg.1 |
NM_003082.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010620.hg.1 |
NM_003082.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010620.hg.1 |
NM_003082.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010620.hg.1 |
NM_003082.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010620.hg.1 |
NM_003082.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010620.hg.1 |
NM_003082.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010620.hg.1 |
NM_003082.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010620.hg.1 |
NM_003082.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010620.hg.1 |
NM_003082.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010620.hg.1 |
NM_003082.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010620.hg.1 |
NM_003082.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010620.hg.1 |
ENST00000216294.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010620.hg.1 |
ENST00000216294.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010620.hg.1 |
ENST00000216294.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010620.hg.1 |
ENST00000216294.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010620.hg.1 |
ENST00000216294.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010620.hg.1 |
ENST00000216294.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010620.hg.1 |
ENST00000216294.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010620.hg.1 |
ENST00000216294.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010620.hg.1 |
ENST00000216294.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010620.hg.1 |
ENST00000216294.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010620.hg.1 |
ENST00000216294.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010620.hg.1 |
ENST00000216294.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010620.hg.1 |
NM_003082.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010620.hg.1 |
NM_003082.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010620.hg.1 |
NM_003082.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010620.hg.1 |
NM_003082.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010620.hg.1 |
NM_003082.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010620.hg.1 |
NM_003082.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010620.hg.1 |
NM_003082.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010620.hg.1 |
NM_003082.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010620.hg.1 |
NM_003082.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010620.hg.1 |
NM_003082.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010620.hg.1 |
NM_003082.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010620.hg.1 |
NM_003082.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400179693.hg.1 |
ENST00000216294.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400179693.hg.1 |
NM_003082.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400179693.hg.1 |
ENST00000216294.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400179693.hg.1 |
NM_003082.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400179693.hg.1 |
TC1400010620.hg.1 |
Clariom D TC |