Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000379.3 | NP_000370.2 | 33 - 34 | 3661 | 62 | |
XM_011533095.2 | XP_011531397.1 | 33 - 34 | 3690 | 62 | ||
GenBank mRNA | D11456.2 | - | 33 - 34 | 3639 | 62 | |
U06117.1 | - | 33 - 34 | 3645 | 62 | ||
U39487.1 | - | 33 - 34 | 3616 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Homo sapiens Probe spans exons |