Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
ENST00000330710.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
ENST00000330710.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
ENST00000330710.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
ENST00000330710.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
ENST00000330710.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
ENST00000330710.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
ENST00000330710.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
ENST00000330710.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
ENST00000330710.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
ENST00000330710.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
ENST00000330710.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
ENST00000330710.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
ENST00000330710.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
ENST00000330710.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
ENST00000357223.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
ENST00000357223.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
ENST00000357223.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
ENST00000357223.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
ENST00000357223.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
ENST00000357223.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
ENST00000357223.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
ENST00000357223.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
ENST00000357223.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
ENST00000357223.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
ENST00000357223.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
ENST00000357223.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
NM_001127258.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
NM_001127258.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
NM_001127258.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
NM_001127258.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
NM_001127258.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
NM_001127258.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
NM_001127258.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
NM_001127258.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
NM_001127258.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
NM_001127258.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
NM_001127258.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
NM_001127258.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
NM_032425.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
NM_032425.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
NM_032425.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
NM_032425.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
NM_032425.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
NM_032425.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
NM_032425.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
NM_032425.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
NM_032425.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
NM_032425.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
NM_032425.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008181.hg.1 |
NM_032425.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400158115.hg.1 |
ENST00000330710.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400158115.hg.1 |
ENST00000357223.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400158115.hg.1 |
NM_001127258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400158115.hg.1 |
NM_032425.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400158115.hg.1 |
ENST00000330710.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400158115.hg.1 |
ENST00000357223.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1400158115.hg.1 |
NM_001127258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400158115.hg.1 |
NM_032425.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1400158115.hg.1 |
TC1400008181.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
ENST00000330710.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
ENST00000330710.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
ENST00000330710.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
ENST00000330710.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
ENST00000330710.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
ENST00000330710.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
ENST00000330710.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
ENST00000330710.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
ENST00000330710.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
ENST00000330710.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
ENST00000330710.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
ENST00000330710.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
ENST00000330710.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
ENST00000330710.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
ENST00000357223.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
ENST00000357223.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
ENST00000357223.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
ENST00000357223.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
ENST00000357223.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
ENST00000357223.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
ENST00000357223.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
ENST00000357223.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
ENST00000357223.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
ENST00000357223.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
ENST00000357223.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
ENST00000357223.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
NM_001127258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
NM_001127258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
NM_001127258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
NM_001127258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
NM_001127258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
NM_001127258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
NM_001127258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
NM_001127258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
NM_001127258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
NM_001127258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
NM_001127258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
NM_001127258.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
NM_032425.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
NM_032425.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
NM_032425.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
NM_032425.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
NM_032425.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
NM_032425.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
NM_032425.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
NM_032425.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
NM_032425.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
NM_032425.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
NM_032425.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400008181.hg.1 |
NM_032425.5 |
Refseq |