Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002286.hg.1 |
TC6_dbb_hap3000125.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3002286.hg.1 |
NM_005275.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011368.hg.1 |
ENST00000376621.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011368.hg.1 |
ENST00000376621.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011368.hg.1 |
ENST00000376621.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011368.hg.1 |
ENST00000376621.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011368.hg.1 |
ENST00000376621.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011368.hg.1 |
ENST00000376621.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011368.hg.1 |
ENST00000376621.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011368.hg.1 |
ENST00000376621.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011368.hg.1 |
ENST00000376621.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011368.hg.1 |
ENST00000376621.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011368.hg.1 |
ENST00000376621.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011368.hg.1 |
ENST00000376621.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011368.hg.1 |
ENST00000376621.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011368.hg.1 |
ENST00000376621.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011368.hg.1 |
ENST00000376621.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011368.hg.1 |
ENST00000376621.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011368.hg.1 |
ENST00000376621.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011368.hg.1 |
ENST00000376621.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011368.hg.1 |
ENST00000376621.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011368.hg.1 |
ENST00000433809.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011368.hg.1 |
ENST00000433809.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011368.hg.1 |
ENST00000433809.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011368.hg.1 |
ENST00000433809.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011368.hg.1 |
ENST00000433809.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011368.hg.1 |
ENST00000433809.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011368.hg.1 |
ENST00000462708.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011368.hg.1 |
ENST00000462708.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011368.hg.1 |
ENST00000462708.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011368.hg.1 |
ENST00000462708.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011368.hg.1 |
ENST00000462708.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011368.hg.1 |
ENST00000462708.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011368.hg.1 |
ENST00000464231.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011368.hg.1 |
ENST00000464231.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011368.hg.1 |
ENST00000464231.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011368.hg.1 |
ENST00000487166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011368.hg.1 |
ENST00000487166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011368.hg.1 |
ENST00000487166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011368.hg.1 |
ENST00000487166.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002130.hg.1 |
TC6_ssto_hap7000117.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7002130.hg.1 |
NM_005275.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001876.hg.1 |
TC6_mann_hap4000114.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001876.hg.1 |
NM_005275.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002329.hg.1 |
TC6_qbl_hap6000126.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6002329.hg.1 |
NM_005275.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016912.hg.1 |
TC06001501.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016912.hg.1 |
ENST00000376621.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016912.hg.1 |
ENST00000429126.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06016912.hg.1 |
NM_005275.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002356.hg.1 |
TC6_cox_hap2000136.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2002356.hg.1 |
NM_005275.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002046.hg.1 |
TC6_mcf_hap5000115.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5002046.hg.1 |
NM_005275.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011368.hg.1 |
ENST00000376621.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011368.hg.1 |
ENST00000376621.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011368.hg.1 |
ENST00000376621.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011368.hg.1 |
ENST00000376621.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011368.hg.1 |
ENST00000376621.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011368.hg.1 |
ENST00000376621.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011368.hg.1 |
ENST00000376621.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011368.hg.1 |
ENST00000376621.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011368.hg.1 |
ENST00000376621.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011368.hg.1 |
ENST00000376621.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011368.hg.1 |
ENST00000376621.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011368.hg.1 |
ENST00000376621.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011368.hg.1 |
ENST00000376621.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011368.hg.1 |
ENST00000376621.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011368.hg.1 |
ENST00000376621.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011368.hg.1 |
ENST00000376621.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011368.hg.1 |
ENST00000376621.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011368.hg.1 |
ENST00000376621.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011368.hg.1 |
ENST00000376621.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011368.hg.1 |
ENST00000433809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011368.hg.1 |
ENST00000433809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011368.hg.1 |
ENST00000433809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011368.hg.1 |
ENST00000433809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011368.hg.1 |
ENST00000433809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011368.hg.1 |
ENST00000433809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011368.hg.1 |
ENST00000462708.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011368.hg.1 |
ENST00000462708.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011368.hg.1 |
ENST00000462708.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011368.hg.1 |
ENST00000462708.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011368.hg.1 |
ENST00000462708.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011368.hg.1 |
ENST00000462708.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011368.hg.1 |
ENST00000464231.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011368.hg.1 |
ENST00000464231.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011368.hg.1 |
ENST00000464231.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011368.hg.1 |
ENST00000487166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011368.hg.1 |
ENST00000487166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011368.hg.1 |
ENST00000487166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011368.hg.1 |
ENST00000487166.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176596.hg.1 |
ENST00000376621.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176596.hg.1 |
ENST00000433809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176596.hg.1 |
ENST00000376621.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176596.hg.1 |
ENST00000433809.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600176596.hg.1 |
TC0600011368.hg.1 |
Clariom D TC |