Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12001711.hg.1 |
ENST00000299665.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12001711.hg.1 |
NM_080387.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12001711.hg.1 |
TC12000130.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006721.hg.1 |
ENST00000299665.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006721.hg.1 |
ENST00000299665.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006721.hg.1 |
ENST00000299665.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006721.hg.1 |
ENST00000299665.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006721.hg.1 |
ENST00000299665.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006721.hg.1 |
ENST00000299665.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006721.hg.1 |
ENST00000299665.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006721.hg.1 |
ENST00000299665.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006721.hg.1 |
ENST00000299665.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006721.hg.1 |
ENST00000299665.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006721.hg.1 |
ENST00000382064.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006721.hg.1 |
ENST00000382064.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006721.hg.1 |
ENST00000382064.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006721.hg.1 |
ENST00000382064.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006721.hg.1 |
ENST00000382064.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006721.hg.1 |
ENST00000382064.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006721.hg.1 |
ENST00000382064.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006721.hg.1 |
NM_080387.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006721.hg.1 |
NM_080387.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006721.hg.1 |
NM_080387.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006721.hg.1 |
NM_080387.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006721.hg.1 |
NM_080387.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006721.hg.1 |
NM_080387.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006721.hg.1 |
NM_080387.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006721.hg.1 |
NM_080387.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006721.hg.1 |
NM_080387.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006721.hg.1 |
NM_080387.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200147039.hg.1 |
ENST00000299665.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200147039.hg.1 |
ENST00000382064.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200147039.hg.1 |
NM_080387.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200147039.hg.1 |
ENST00000299665.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200147039.hg.1 |
ENST00000382064.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200147039.hg.1 |
NM_080387.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200147039.hg.1 |
TC1200006721.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200006721.hg.1 |
ENST00000299665.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006721.hg.1 |
ENST00000299665.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006721.hg.1 |
ENST00000299665.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006721.hg.1 |
ENST00000299665.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006721.hg.1 |
ENST00000299665.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006721.hg.1 |
ENST00000299665.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006721.hg.1 |
ENST00000299665.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006721.hg.1 |
ENST00000299665.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006721.hg.1 |
ENST00000299665.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006721.hg.1 |
ENST00000299665.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006721.hg.1 |
ENST00000382064.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006721.hg.1 |
ENST00000382064.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006721.hg.1 |
ENST00000382064.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006721.hg.1 |
ENST00000382064.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006721.hg.1 |
ENST00000382064.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006721.hg.1 |
ENST00000382064.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006721.hg.1 |
ENST00000382064.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006721.hg.1 |
NM_080387.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006721.hg.1 |
NM_080387.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006721.hg.1 |
NM_080387.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006721.hg.1 |
NM_080387.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006721.hg.1 |
NM_080387.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006721.hg.1 |
NM_080387.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006721.hg.1 |
NM_080387.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006721.hg.1 |
NM_080387.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006721.hg.1 |
NM_080387.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006721.hg.1 |
NM_080387.4 |
Refseq |