Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPB41L4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK025249.1 | - | 1 | 3051 | 152 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ferric chelate reductase 1 like | FRRS1L |
C9orf4, CG-6, CG6, EIEE37 |
-82.2kb | Anti-sense | Chr. 9 - 109136188 - 109167291 on NCBI Build GRCh38 |
Downstream |
protein tyrosine phosphatase, non-receptor type 3 |
PTPN3 |
PTP-H1, PTPH1 |
125.7kb | Anti-sense | Chr. 9 - 109375694 - 109538395 on NCBI Build GRCh38 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon |