Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002470.3 | NP_002461.2 | 11 - 12 | 1094 | 100 | |
XM_011523870.2 | XP_011522172.1 | 11 - 12 | 2461 | 100 | ||
XM_011523871.2 | XP_011522173.1 | 11 - 12 | 1094 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Probe spans exons |