Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
ENST00000324361.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
ENST00000324361.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
ENST00000324361.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
ENST00000324361.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
ENST00000324361.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
ENST00000324361.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
ENST00000324361.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
ENST00000324361.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
ENST00000324361.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
ENST00000324361.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
ENST00000397621.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
ENST00000397621.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
ENST00000397621.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
ENST00000397621.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
ENST00000397621.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
ENST00000397621.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
ENST00000397621.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
ENST00000397621.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
ENST00000397621.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
ENST00000397621.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
ENST00000397621.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
ENST00000562563.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
ENST00000562563.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
ENST00000562563.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
ENST00000562563.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
ENST00000562563.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
ENST00000562563.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
ENST00000562563.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
ENST00000562585.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
ENST00000562585.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
ENST00000562585.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
ENST00000562648.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
ENST00000562648.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
ENST00000562648.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
ENST00000562648.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
ENST00000562648.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
ENST00000562648.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
ENST00000562648.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
ENST00000562648.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
ENST00000562648.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
NM_153350.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
NM_153350.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
NM_153350.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
NM_153350.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
NM_153350.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
NM_153350.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
NM_153350.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
NM_153350.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
NM_153350.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
NM_153350.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008973.hg.1 |
NM_153350.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600167760.hg.1 |
ENST00000324361.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600167760.hg.1 |
ENST00000397621.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600167760.hg.1 |
NM_153350.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600167760.hg.1 |
ENST00000324361.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600167760.hg.1 |
ENST00000397621.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600167760.hg.1 |
NM_153350.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600167760.hg.1 |
TC1600008973.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600167762.hg.1 |
ENST00000324361.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600167762.hg.1 |
ENST00000397621.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600167762.hg.1 |
NM_153350.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600167762.hg.1 |
ENST00000324361.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600167762.hg.1 |
ENST00000397621.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600167762.hg.1 |
NM_153350.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600167762.hg.1 |
TC1600008973.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
ENST00000324361.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
ENST00000324361.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
ENST00000324361.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
ENST00000324361.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
ENST00000324361.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
ENST00000324361.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
ENST00000324361.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
ENST00000324361.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
ENST00000324361.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
ENST00000324361.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
ENST00000397621.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
ENST00000397621.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
ENST00000397621.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
ENST00000397621.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
ENST00000397621.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
ENST00000397621.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
ENST00000397621.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
ENST00000397621.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
ENST00000397621.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
ENST00000397621.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
ENST00000397621.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
ENST00000562563.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
ENST00000562563.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
ENST00000562563.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
ENST00000562563.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
ENST00000562563.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
ENST00000562563.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
ENST00000562563.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
ENST00000562585.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
ENST00000562585.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
ENST00000562585.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
ENST00000562648.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
ENST00000562648.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
ENST00000562648.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
ENST00000562648.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
ENST00000562648.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
ENST00000562648.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
ENST00000562648.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
ENST00000562648.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
ENST00000562648.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
NM_153350.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
NM_153350.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
NM_153350.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
NM_153350.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
NM_153350.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
NM_153350.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
NM_153350.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
NM_153350.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
NM_153350.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
NM_153350.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008973.hg.1 |
NM_153350.3 |
Refseq |