Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MORC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014429.3 | NP_055244.3 | 26 - 27 | 2687 | 159 | |
XM_005247362.2 | XP_005247419.1 | 27 - 28 | 2734 | 159 | ||
XM_011512691.1 | XP_011510993.1 | 27 - 28 | 2740 | 159 | ||
XM_011512692.1 | XP_011510994.1 | 26 - 27 | 2650 | 159 | ||
XM_011512694.1 | XP_011510996.1 | 24 - 25 | 2491 | 159 | ||
XM_011512696.2 | XP_011510998.1 | 21 - 22 | 2191 | 159 | ||
XM_011512697.2 | XP_011510999.1 | 20 - 21 | 2359 | 159 | ||
XM_017006169.1 | XP_016861658.1 | 27 - 28 | 2740 | 159 | ||
XM_017006170.1 | XP_016861659.1 | 19 - 20 | 2069 | 159 | ||
GenBank mRNA | AF084946.1 | - | 26 - 27 | 2662 | 159 | |
AK302642.1 | - | 25 - 26 | 2581 | 159 | ||
AL353955.1 | - | 20 - 21 | 2047 | 159 | ||
BC028680.1 | - | 26 - 27 | 2688 | 159 | ||
BC050307.1 | - | 26 - 27 | 2626 | 4838254 | 159 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons |