Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDEM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK293583.1 | - | 4 - 5 | 624 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Probe spans exons |