Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000258484.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000258484.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000258484.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000258484.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000258484.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000258484.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000258484.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000258484.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000258484.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000258484.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000258484.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000258484.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000258484.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000258484.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000258484.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000258484.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000258484.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000258484.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000258484.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000397424.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000397424.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000397424.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000397424.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000397424.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000397424.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000409654.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000409654.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000409654.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000409654.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000409654.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000409654.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000449013.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000449013.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000457184.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000457184.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000457184.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000457184.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000457184.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000457184.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000491099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000491099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000491099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000491099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000491099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
ENST00000491099.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
NM_015630.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
NM_015630.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
NM_015630.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
NM_015630.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
NM_015630.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
NM_015630.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
NM_015630.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
NM_015630.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
NM_015630.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
NM_015630.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
NM_015630.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
NM_015630.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
NM_015630.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
NM_015630.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
NM_015630.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
NM_015630.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
NM_015630.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
NM_015630.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200009612.hg.1 |
NM_015630.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02013523.hg.1 |
TC02000926.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02013523.hg.1 |
ENST00000258484.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02013523.hg.1 |
NM_015630.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000258484.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000258484.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000258484.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000258484.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000258484.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000258484.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000258484.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000258484.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000258484.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000258484.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000258484.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000258484.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000258484.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000258484.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000258484.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000258484.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000258484.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000258484.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000258484.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000397424.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000397424.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000397424.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000397424.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000397424.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000397424.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000409654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000409654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000409654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000409654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000409654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000409654.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000449013.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000449013.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000457184.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000457184.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000457184.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000457184.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000457184.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000457184.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000491099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000491099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000491099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000491099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000491099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
ENST00000491099.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
NM_015630.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
NM_015630.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
NM_015630.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
NM_015630.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
NM_015630.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
NM_015630.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
NM_015630.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
NM_015630.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
NM_015630.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
NM_015630.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
NM_015630.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
NM_015630.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
NM_015630.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
NM_015630.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
NM_015630.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
NM_015630.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
NM_015630.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
NM_015630.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200009612.hg.1 |
NM_015630.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200167206.hg.1 |
ENST00000258484.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200167206.hg.1 |
NM_015630.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200167206.hg.1 |
ENST00000258484.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200167206.hg.1 |
NM_015630.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200167206.hg.1 |
TC0200009612.hg.1 |
Clariom D TC |