Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000466175.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000466175.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000466175.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000466175.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000466175.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000466175.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000466175.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000505551.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000505551.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000505551.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000505551.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000505551.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000505551.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000505551.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000510940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000510940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000510940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000511977.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000511977.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000511977.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000511977.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000511977.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000512189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000512189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000512189.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000512669.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000512669.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000512669.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
ENST00000512669.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04013247.hg.1 |
TC04000970.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04013247.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04013247.hg.1 |
ENST00000417150.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04013247.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000466175.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000466175.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000466175.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000466175.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000466175.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000466175.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000466175.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000505551.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000505551.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000505551.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000505551.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000505551.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000505551.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000505551.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000510940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000510940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000510940.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000511977.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000511977.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000511977.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000511977.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000511977.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000512189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000512189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000512189.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000512669.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000512669.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000512669.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
ENST00000512669.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400009754.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400166968.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400166968.hg.1 |
ENST00000511977.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400166968.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400166968.hg.1 |
ENST00000302787.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400166968.hg.1 |
ENST00000511977.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400166968.hg.1 |
NM_012318.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400166968.hg.1 |
TC0400009754.hg.1 |
Clariom D TC |