Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004491.hg.1 |
TC08000318.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004491.hg.1 |
ENST00000276533.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004491.hg.1 |
ENST00000518671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004491.hg.1 |
ENST00000520710.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004491.hg.1 |
ENST00000523277.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004491.hg.1 |
NM_032336.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000276533.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000276533.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000276533.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000276533.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000276533.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000276533.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000276533.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000276533.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000276533.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000276533.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000276533.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000276533.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000276533.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000518430.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000518430.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000518430.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000518430.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000518671.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000518671.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000518671.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000518671.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000518671.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000518671.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000518671.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000518671.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000518671.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000518671.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000520354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000520354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000520354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000520354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000520354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000520354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000520354.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000520631.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000520631.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000520631.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000520631.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000520710.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000520710.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000520710.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000520710.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000520710.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000520710.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000520736.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000520736.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000520736.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000522642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000522642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000522642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000522642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000523277.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000523277.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000523277.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000523277.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000523277.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000523277.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000523277.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000523277.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000523277.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000523277.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000523277.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000523277.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000523277.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
ENST00000523277.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
NM_032336.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
NM_032336.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
NM_032336.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
NM_032336.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
NM_032336.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
NM_032336.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
NM_032336.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
NM_032336.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
NM_032336.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
NM_032336.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
NM_032336.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
NM_032336.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007416.hg.1 |
NM_032336.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004494.hg.1 |
TC08000318.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004494.hg.1 |
ENST00000276533.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004494.hg.1 |
ENST00000518671.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004494.hg.1 |
ENST00000520710.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004494.hg.1 |
ENST00000523277.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004494.hg.1 |
NM_032336.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800151151.hg.1 |
ENST00000276533.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151151.hg.1 |
ENST00000518671.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151151.hg.1 |
ENST00000520354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151151.hg.1 |
ENST00000520631.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151151.hg.1 |
ENST00000520710.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151151.hg.1 |
ENST00000520736.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151151.hg.1 |
ENST00000523277.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151151.hg.1 |
NM_032336.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800151151.hg.1 |
ENST00000276533.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151151.hg.1 |
ENST00000518671.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151151.hg.1 |
ENST00000520354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151151.hg.1 |
ENST00000520631.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151151.hg.1 |
ENST00000520710.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151151.hg.1 |
ENST00000520736.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151151.hg.1 |
ENST00000523277.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151151.hg.1 |
NM_032336.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800151151.hg.1 |
TC0800007416.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000276533.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000276533.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000276533.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000276533.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000276533.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000276533.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000276533.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000276533.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000276533.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000276533.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000276533.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000276533.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000276533.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000518430.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000518430.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000518430.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000518430.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000518671.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000518671.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000518671.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000518671.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000518671.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000518671.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000518671.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000518671.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000518671.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000518671.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000520354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000520354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000520354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000520354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000520354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000520354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000520354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000520631.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000520631.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000520631.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000520631.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000520710.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000520710.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000520710.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000520710.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000520710.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000520710.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000520736.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000520736.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000520736.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000522642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000522642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000522642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000522642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000523277.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000523277.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000523277.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000523277.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000523277.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000523277.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000523277.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000523277.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000523277.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000523277.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000523277.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000523277.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000523277.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
ENST00000523277.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
NM_032336.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
NM_032336.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
NM_032336.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
NM_032336.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
NM_032336.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
NM_032336.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
NM_032336.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
NM_032336.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
NM_032336.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
NM_032336.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
NM_032336.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
NM_032336.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007416.hg.1 |
NM_032336.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800151155.hg.1 |
ENST00000276533.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151155.hg.1 |
ENST00000518671.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151155.hg.1 |
ENST00000520354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151155.hg.1 |
ENST00000520631.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151155.hg.1 |
ENST00000520710.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151155.hg.1 |
ENST00000520736.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151155.hg.1 |
ENST00000523277.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151155.hg.1 |
NM_032336.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800151155.hg.1 |
ENST00000276533.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151155.hg.1 |
ENST00000518671.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151155.hg.1 |
ENST00000520354.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151155.hg.1 |
ENST00000520631.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151155.hg.1 |
ENST00000520710.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151155.hg.1 |
ENST00000520736.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151155.hg.1 |
ENST00000523277.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151155.hg.1 |
NM_032336.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800151155.hg.1 |
TC0800007416.hg.1 |
Clariom D TC |