Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000467505.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000467505.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000467505.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000467505.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000480491.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000480491.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000480491.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000484879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000484879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000484879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000484879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000484879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000484879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000484879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000525036.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000525036.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000525036.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000525036.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000525036.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000525036.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000525036.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000525036.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000525036.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000531171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000531171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000531171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000531171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000531171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
ENST00000531171.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002192.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002192.hg.1 |
ENST00000542917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002192.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002192.hg.1 |
TC13000190.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002195.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002195.hg.1 |
ENST00000542917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002195.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002195.hg.1 |
TC13000190.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002193.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002193.hg.1 |
ENST00000542917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002193.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002193.hg.1 |
TC13000190.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002194.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002194.hg.1 |
ENST00000542917.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002194.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13002194.hg.1 |
TC13000190.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1300148351.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300148351.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300148351.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300148351.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300148351.hg.1 |
TC1300007137.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300148353.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300148353.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300148353.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300148353.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300148353.hg.1 |
TC1300007137.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300148352.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300148352.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300148352.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300148352.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300148352.hg.1 |
TC1300007137.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300148350.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300148350.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300148350.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300148350.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300148350.hg.1 |
TC1300007137.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000267163.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000467505.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000467505.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000467505.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000467505.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000480491.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000480491.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000480491.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000484879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000484879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000484879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000484879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000484879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000484879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000484879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000525036.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000525036.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000525036.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000525036.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000525036.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000525036.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000525036.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000525036.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000525036.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000531171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000531171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000531171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000531171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000531171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
ENST00000531171.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300007137.hg.1 |
NM_000321.2 |
Refseq |