Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000238497.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000238497.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000238497.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000238497.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000238497.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000238497.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000238497.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000238497.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000238497.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000238497.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000238497.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000238497.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000238497.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000238497.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000238497.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000238497.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000238497.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000238497.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000238497.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000588059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000588059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000588059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000588059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000588059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000588059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000588059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000588059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000588059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000588059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000588059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000588059.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000588323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000588323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000588323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000588323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000588323.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000589604.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000589604.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000589604.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000589604.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000589604.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000589604.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000589604.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000589604.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000589604.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000589604.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000589604.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000589604.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000589604.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000591383.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000591383.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000591383.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000591383.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000591383.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000591475.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000591475.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000591475.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000591475.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000591475.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000591475.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000591475.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000591475.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000591475.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000591475.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000591475.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000591475.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000591475.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000591475.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000591475.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000591519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000591519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000591519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000591519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000591519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
ENST00000591519.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
NM_004869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
NM_004869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
NM_004869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
NM_004869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
NM_004869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
NM_004869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
NM_004869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
NM_004869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
NM_004869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
NM_004869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
NM_004869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
NM_004869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
NM_004869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
NM_004869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
NM_004869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
NM_004869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
NM_004869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
NM_004869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
NM_004869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800008906.hg.1 |
NM_004869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18005794.hg.1 |
ENST00000238497.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18005794.hg.1 |
NM_004869.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18005794.hg.1 |
TC18000557.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1800161190.hg.1 |
ENST00000238497.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800161190.hg.1 |
ENST00000588059.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800161190.hg.1 |
ENST00000589604.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800161190.hg.1 |
ENST00000591383.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800161190.hg.1 |
ENST00000591475.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800161190.hg.1 |
NM_004869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800161190.hg.1 |
TC1800008906.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800161190.hg.1 |
ENST00000238497.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800161190.hg.1 |
ENST00000588059.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800161190.hg.1 |
ENST00000589604.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800161190.hg.1 |
ENST00000591383.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800161190.hg.1 |
ENST00000591475.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800161190.hg.1 |
NM_004869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000238497.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000238497.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000238497.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000238497.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000238497.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000238497.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000238497.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000238497.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000238497.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000238497.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000238497.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000238497.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000238497.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000238497.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000238497.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000238497.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000238497.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000238497.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000238497.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000588059.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000588059.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000588059.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000588059.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000588059.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000588059.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000588059.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000588059.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000588059.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000588059.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000588059.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000588059.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000588323.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000588323.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000588323.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000588323.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000588323.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000589604.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000589604.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000589604.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000589604.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000589604.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000589604.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000589604.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000589604.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000589604.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000589604.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000589604.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000589604.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000589604.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000591383.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000591383.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000591383.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000591383.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000591383.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000591475.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000591475.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000591475.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000591475.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000591475.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000591475.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000591475.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000591475.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000591475.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000591475.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000591475.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000591475.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000591475.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000591475.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000591475.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000591519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000591519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000591519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000591519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000591519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
ENST00000591519.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
NM_004869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
NM_004869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
NM_004869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
NM_004869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
NM_004869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
NM_004869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
NM_004869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
NM_004869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
NM_004869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
NM_004869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
NM_004869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
NM_004869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
NM_004869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
NM_004869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
NM_004869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
NM_004869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
NM_004869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
NM_004869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
NM_004869.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800008906.hg.1 |
NM_004869.3 |
Refseq |