Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291860.1 | NP_001278789.1 | 85 - 86 | 11750 | 85 | |
NM_005529.6 | NP_005520.4 | 85 - 86 | 11747 | 85 | ||
XM_011541318.2 | XP_011539620.1 | 90 - 91 | 12231 | 85 | ||
XM_017001120.1 | XP_016856609.1 | 86 - 87 | 11877 | 85 | ||
XM_017001121.1 | XP_016856610.1 | 85 - 86 | 11826 | 85 | ||
XM_017001122.1 | XP_016856611.1 | 85 - 86 | 11823 | 85 | ||
GenBank mRNA | AB209851.1 | - | 37 - 38 | 5487 | 85 | |
BC109204.1 | - | 4 - 5 | 373 | 85 | ||
M85289.1 | - | 85 - 86 | 11747 | 85 | ||
X62515.1 | - | 85 - 86 | 11713 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |