Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKD1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138295.3 | NP_612152.1 | 33 - 34 | 5272 | 119 | |
XM_017011798.1 | XP_016867287.1 | 34 - 35 | 11677 | 119 | ||
GenBank mRNA | AB061683.1 | - | 33 - 34 | 5238 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Homo sapiens Probe spans exons |