Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001308152.1 | NP_001295081.1 | 22 - 23 | 2763 | 128 | |
NM_001308153.1 | NP_001295082.1 | 24 - 25 | 2966 | 128 | ||
NM_014629.3 | NP_055444.2 | 23 - 24 | 2877 | 128 | ||
XM_005266041.3 | XP_005266098.1 | 23 - 24 | 2819 | 128 | ||
XM_011534767.2 | XP_011533069.1 | 21 - 22 | 2622 | 128 | ||
XM_011534768.1 | XP_011533070.1 | 22 - 23 | 2742 | 128 | ||
XM_011534770.2 | XP_011533072.1 | 22 - 23 | 2742 | 128 | ||
XM_017014003.1 | XP_016869492.1 | 23 - 24 | 2782 | 128 | ||
GenBank mRNA | AB002292.2 | - | 23 - 24 | 2876 | 128 | |
BC112926.1 | - | 22 - 23 | 2751 | 6500770 | 128 | |
CR749570.1 | - | 24 - 25 | 2927 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |