Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017019084.1 | XP_016874573.1 | 2 - 3 | 59 | 95 | |
XM_017019085.1 | XP_016874574.1 | 3 - 4 | 339 | 95 | ||
XM_017019086.1 | XP_016874575.1 | 3 - 4 | 344 | 95 | ||
GenBank mRNA | AB023201.1 | - | 2 - 3 | 53 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Homo sapiens Probe spans exons |