Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004309.hg.1 |
ENST00000269137.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004309.hg.1 |
ENST00000269138.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004309.hg.1 |
ENST00000415083.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004309.hg.1 |
ENST00000539244.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004309.hg.1 |
ENST00000539849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004309.hg.1 |
ENST00000542420.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004309.hg.1 |
ENST00000542743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004309.hg.1 |
ENST00000545952.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004309.hg.1 |
NM_001007559.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004309.hg.1 |
NM_005637.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004309.hg.1 |
TC18000423.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004310.hg.1 |
ENST00000269137.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004310.hg.1 |
ENST00000269138.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004310.hg.1 |
ENST00000415083.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004310.hg.1 |
ENST00000539244.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004310.hg.1 |
ENST00000539849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004310.hg.1 |
ENST00000542420.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004310.hg.1 |
ENST00000542743.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004310.hg.1 |
ENST00000545952.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004310.hg.1 |
NM_001007559.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004310.hg.1 |
NM_005637.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18004310.hg.1 |
TC18000423.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1800156786.hg.1 |
ENST00000269137.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156786.hg.1 |
ENST00000269138.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156786.hg.1 |
ENST00000415083.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156786.hg.1 |
ENST00000542420.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156786.hg.1 |
ENST00000579640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156786.hg.1 |
ENST00000580958.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156786.hg.1 |
ENST00000585121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156786.hg.1 |
NM_001007559.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156786.hg.1 |
NM_001308201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156786.hg.1 |
NM_005637.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156786.hg.1 |
TC1800008331.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800156786.hg.1 |
ENST00000269137.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156786.hg.1 |
ENST00000269138.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156786.hg.1 |
ENST00000415083.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156786.hg.1 |
ENST00000542420.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156786.hg.1 |
ENST00000579640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156786.hg.1 |
ENST00000580958.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156786.hg.1 |
ENST00000585121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156786.hg.1 |
NM_001007559.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156786.hg.1 |
NM_001308201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156786.hg.1 |
NM_005637.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156783.hg.1 |
ENST00000269137.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156783.hg.1 |
ENST00000269138.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156783.hg.1 |
ENST00000415083.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156783.hg.1 |
ENST00000542420.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156783.hg.1 |
ENST00000579640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156783.hg.1 |
ENST00000580958.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156783.hg.1 |
ENST00000582092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156783.hg.1 |
ENST00000585121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156783.hg.1 |
NM_001007559.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156783.hg.1 |
NM_001308201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156783.hg.1 |
NM_005637.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156783.hg.1 |
TC1800008331.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800156783.hg.1 |
ENST00000269137.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156783.hg.1 |
ENST00000269138.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156783.hg.1 |
ENST00000415083.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156783.hg.1 |
ENST00000542420.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156783.hg.1 |
ENST00000579640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156783.hg.1 |
ENST00000580958.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156783.hg.1 |
ENST00000582092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156783.hg.1 |
ENST00000585121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156783.hg.1 |
NM_001007559.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156783.hg.1 |
NM_001308201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156783.hg.1 |
NM_005637.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156785.hg.1 |
ENST00000269137.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156785.hg.1 |
ENST00000269138.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156785.hg.1 |
ENST00000415083.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156785.hg.1 |
ENST00000542420.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156785.hg.1 |
ENST00000579640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156785.hg.1 |
ENST00000580958.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156785.hg.1 |
ENST00000582092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156785.hg.1 |
ENST00000585121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156785.hg.1 |
NM_001007559.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156785.hg.1 |
NM_001308201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156785.hg.1 |
NM_005637.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156785.hg.1 |
TC1800008331.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800156785.hg.1 |
ENST00000269137.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156785.hg.1 |
ENST00000269138.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156785.hg.1 |
ENST00000415083.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156785.hg.1 |
ENST00000542420.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156785.hg.1 |
ENST00000579640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156785.hg.1 |
ENST00000580958.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156785.hg.1 |
ENST00000582092.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156785.hg.1 |
ENST00000585121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156785.hg.1 |
NM_001007559.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156785.hg.1 |
NM_001308201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156785.hg.1 |
NM_005637.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156787.hg.1 |
ENST00000269137.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156787.hg.1 |
ENST00000269138.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156787.hg.1 |
ENST00000415083.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156787.hg.1 |
ENST00000539244.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156787.hg.1 |
ENST00000542420.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156787.hg.1 |
ENST00000579640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156787.hg.1 |
ENST00000580958.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156787.hg.1 |
ENST00000585121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156787.hg.1 |
NM_001007559.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156787.hg.1 |
NM_001308201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156787.hg.1 |
NM_005637.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156787.hg.1 |
TC1800008331.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800156787.hg.1 |
ENST00000269137.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156787.hg.1 |
ENST00000269138.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156787.hg.1 |
ENST00000415083.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156787.hg.1 |
ENST00000539244.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156787.hg.1 |
ENST00000542420.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156787.hg.1 |
ENST00000579640.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156787.hg.1 |
ENST00000580958.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156787.hg.1 |
ENST00000585121.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800156787.hg.1 |
NM_001007559.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156787.hg.1 |
NM_001308201.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800156787.hg.1 |
NM_005637.3 |
Refseq |