Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002858.3 | NP_002849.1 | 18 - 19 | 1632 | 82 | |
XM_006710802.2 | XP_006710865.2 | 19 - 20 | 1995 | 82 | ||
XM_011541877.1 | XP_011540179.1 | 11 - 12 | 878 | 82 | ||
XM_017001952.1 | XP_016857441.1 | 18 - 19 | 2173 | 82 | ||
XM_017001953.1 | XP_016857442.1 | 18 - 19 | 1553 | 82 | ||
XM_017001954.1 | XP_016857443.1 | 17 - 18 | 1613 | 82 | ||
GenBank mRNA | AK296791.1 | - | 19 - 20 | 1668 | 82 | |
AK302714.1 | - | 17 - 18 | 1557 | 82 | ||
BC068509.1 | - | 14 - 15 | 1263 | 5265201 | 82 | |
BX648715.1 | - | 9 - 10 | 2380 | 82 | ||
M81182.1 | - | 18 - 19 | 1530 | 82 | ||
X58528.1 | - | 18 - 19 | 1554 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |