Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20003293.hg.1 |
ENST00000246186.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20003293.hg.1 |
ENST00000540655.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20003293.hg.1 |
NM_006690.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20003293.hg.1 |
TC20000250.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007213.hg.1 |
ENST00000246186.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007213.hg.1 |
ENST00000246186.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007213.hg.1 |
ENST00000246186.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007213.hg.1 |
ENST00000246186.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007213.hg.1 |
ENST00000246186.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007213.hg.1 |
ENST00000246186.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007213.hg.1 |
ENST00000246186.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007213.hg.1 |
ENST00000246186.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007213.hg.1 |
ENST00000246186.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007213.hg.1 |
ENST00000246186.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007213.hg.1 |
ENST00000246186.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007213.hg.1 |
NM_006690.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007213.hg.1 |
NM_006690.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007213.hg.1 |
NM_006690.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007213.hg.1 |
NM_006690.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007213.hg.1 |
NM_006690.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007213.hg.1 |
NM_006690.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007213.hg.1 |
NM_006690.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007213.hg.1 |
NM_006690.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007213.hg.1 |
NM_006690.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007213.hg.1 |
NM_006690.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000149576.hg.1 |
ENST00000246186.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149576.hg.1 |
NM_006690.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000149576.hg.1 |
TC2000007213.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000149576.hg.1 |
ENST00000246186.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000149576.hg.1 |
NM_006690.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007213.hg.1 |
ENST00000246186.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007213.hg.1 |
ENST00000246186.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007213.hg.1 |
ENST00000246186.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007213.hg.1 |
ENST00000246186.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007213.hg.1 |
ENST00000246186.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007213.hg.1 |
ENST00000246186.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007213.hg.1 |
ENST00000246186.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007213.hg.1 |
ENST00000246186.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007213.hg.1 |
ENST00000246186.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007213.hg.1 |
ENST00000246186.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007213.hg.1 |
ENST00000246186.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007213.hg.1 |
NM_006690.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007213.hg.1 |
NM_006690.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007213.hg.1 |
NM_006690.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007213.hg.1 |
NM_006690.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007213.hg.1 |
NM_006690.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007213.hg.1 |
NM_006690.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007213.hg.1 |
NM_006690.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007213.hg.1 |
NM_006690.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007213.hg.1 |
NM_006690.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007213.hg.1 |
NM_006690.3 |
Refseq |