Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373206.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373206.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373206.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373206.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373206.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373206.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373206.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373206.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373206.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373206.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373206.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373206.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373206.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373206.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373206.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373206.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373206.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373206.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373206.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000373206.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
ENST00000624357.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01007142.hg.1 |
TC01000463.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01007142.hg.1 |
ENST00000373204.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01007142.hg.1 |
ENST00000373206.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01007142.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373206.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373206.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373206.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373206.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373206.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373206.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373206.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373206.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373206.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373206.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373206.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373206.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373206.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373206.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373206.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373206.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373206.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373206.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373206.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000373206.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
ENST00000624357.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100007786.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100154818.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154818.hg.1 |
ENST00000373206.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154818.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100154818.hg.1 |
ENST00000373204.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154818.hg.1 |
ENST00000373206.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100154818.hg.1 |
NM_012199.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100154818.hg.1 |
TC0100007786.hg.1 |
Clariom D TC |