Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012470.hg.1 |
ENST00000278947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012470.hg.1 |
ENST00000278947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012470.hg.1 |
ENST00000278947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012470.hg.1 |
ENST00000278947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012470.hg.1 |
ENST00000278947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012470.hg.1 |
ENST00000278947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012470.hg.1 |
ENST00000278947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012470.hg.1 |
ENST00000278947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012470.hg.1 |
ENST00000278947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012470.hg.1 |
NM_004588.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012470.hg.1 |
NM_004588.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012470.hg.1 |
NM_004588.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012470.hg.1 |
NM_004588.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012470.hg.1 |
NM_004588.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012470.hg.1 |
NM_004588.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012470.hg.1 |
NM_004588.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11028113.hg.1 |
ENST00000278947.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11028113.hg.1 |
NM_004588.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11028113.hg.1 |
TC11002343.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100195861.hg.1 |
ENST00000278947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195861.hg.1 |
NM_004588.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195861.hg.1 |
ENST00000278947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100195861.hg.1 |
NM_004588.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100195861.hg.1 |
TC1100012470.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100012470.hg.1 |
ENST00000278947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012470.hg.1 |
ENST00000278947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012470.hg.1 |
ENST00000278947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012470.hg.1 |
ENST00000278947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012470.hg.1 |
ENST00000278947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012470.hg.1 |
ENST00000278947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012470.hg.1 |
ENST00000278947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012470.hg.1 |
ENST00000278947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012470.hg.1 |
ENST00000278947.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012470.hg.1 |
NM_004588.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012470.hg.1 |
NM_004588.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012470.hg.1 |
NM_004588.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012470.hg.1 |
NM_004588.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012470.hg.1 |
NM_004588.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012470.hg.1 |
NM_004588.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012470.hg.1 |
NM_004588.4 |
Refseq |