Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
ENST00000233710.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
ENST00000233710.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
ENST00000233710.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
ENST00000233710.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
ENST00000233710.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
ENST00000233710.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
ENST00000233710.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
ENST00000233710.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
ENST00000233710.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
ENST00000233710.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
ENST00000233710.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
ENST00000233710.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
ENST00000233710.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
ENST00000233710.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
ENST00000233710.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
ENST00000233710.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
ENST00000233710.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
ENST00000482502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
ENST00000482502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
ENST00000482502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
ENST00000482502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
ENST00000482523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
ENST00000482523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
ENST00000482523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
ENST00000498120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
ENST00000498120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
ENST00000498120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
ENST00000498120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
ENST00000498120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
ENST00000498120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
NM_001608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
NM_001608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
NM_001608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
NM_001608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
NM_001608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
NM_001608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
NM_001608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
NM_001608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
NM_001608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
NM_001608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
NM_001608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
NM_001608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
NM_001608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
NM_001608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
NM_001608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015603.hg.1 |
NM_001608.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043848.hg.1 |
TC02002731.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043848.hg.1 |
ENST00000233710.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02043848.hg.1 |
NM_001608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200214442.hg.1 |
ENST00000233710.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200214442.hg.1 |
NM_001608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200214442.hg.1 |
ENST00000233710.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200214442.hg.1 |
NM_001608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200214442.hg.1 |
TC0200015603.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
ENST00000233710.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
ENST00000233710.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
ENST00000233710.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
ENST00000233710.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
ENST00000233710.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
ENST00000233710.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
ENST00000233710.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
ENST00000233710.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
ENST00000233710.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
ENST00000233710.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
ENST00000233710.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
ENST00000233710.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
ENST00000233710.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
ENST00000233710.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
ENST00000233710.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
ENST00000233710.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
ENST00000233710.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
ENST00000482502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
ENST00000482502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
ENST00000482502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
ENST00000482502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
ENST00000482523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
ENST00000482523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
ENST00000482523.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
ENST00000498120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
ENST00000498120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
ENST00000498120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
ENST00000498120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
ENST00000498120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
ENST00000498120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
NM_001608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
NM_001608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
NM_001608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
NM_001608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
NM_001608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
NM_001608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
NM_001608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
NM_001608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
NM_001608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
NM_001608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
NM_001608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
NM_001608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
NM_001608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
NM_001608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
NM_001608.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015603.hg.1 |
NM_001608.3 |
Refseq |