Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020745.3 | NP_065796.1 | 20 - 21 | 2682 | 63 | |
XM_005249245.3 | XP_005249302.1 | 18 - 19 | 2423 | 63 | ||
XM_011514764.2 | XP_011513066.1 | 20 - 21 | 2714 | 63 | ||
XM_017011112.1 | XP_016866601.1 | 18 - 19 | 2264 | 63 | ||
GenBank mRNA | AB033096.1 | - | 20 - 21 | 2682 | 63 | |
AL536467.3 | - | 3 - 4 | 279 | 63 | ||
BC013593.1 | - | 11 - 12 | 1377 | 63 | ||
BC033169.1 | - | 17 - 18 | 2079 | 63 | ||
BC128175.1 | - | 3 - 4 | 238 | 63 | ||
BC131728.1 | - | 20 - 21 | 2682 | 40124256 | 63 | |
BF768723.1 | - | 1 - 2 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |