Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X012872.hg.1 |
TC0X000965.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X012872.hg.1 |
ENST00000327877.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X012872.hg.1 |
ENST00000378421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X012872.hg.1 |
ENST00000378426.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X012872.hg.1 |
ENST00000440784.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X012872.hg.1 |
NM_144970.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009440.hg.1 |
ENST00000327877.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009440.hg.1 |
ENST00000327877.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009440.hg.1 |
ENST00000327877.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009440.hg.1 |
ENST00000327877.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009440.hg.1 |
ENST00000327877.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009440.hg.1 |
ENST00000327877.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009440.hg.1 |
ENST00000327877.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009440.hg.1 |
ENST00000327877.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009440.hg.1 |
ENST00000327877.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009440.hg.1 |
ENST00000378421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009440.hg.1 |
ENST00000378421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009440.hg.1 |
ENST00000378421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009440.hg.1 |
ENST00000378421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009440.hg.1 |
ENST00000378421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009440.hg.1 |
ENST00000378421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009440.hg.1 |
ENST00000378421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009440.hg.1 |
ENST00000378421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009440.hg.1 |
ENST00000378421.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009440.hg.1 |
ENST00000378426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009440.hg.1 |
ENST00000378426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009440.hg.1 |
ENST00000378426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009440.hg.1 |
ENST00000378426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009440.hg.1 |
ENST00000378426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009440.hg.1 |
ENST00000378426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009440.hg.1 |
ENST00000378426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009440.hg.1 |
ENST00000378426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009440.hg.1 |
ENST00000378426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009440.hg.1 |
ENST00000378426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009440.hg.1 |
ENST00000378426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009440.hg.1 |
ENST00000378426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009440.hg.1 |
ENST00000378426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009440.hg.1 |
NM_144970.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009440.hg.1 |
NM_144970.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009440.hg.1 |
NM_144970.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009440.hg.1 |
NM_144970.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009440.hg.1 |
NM_144970.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009440.hg.1 |
NM_144970.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009440.hg.1 |
NM_144970.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009440.hg.1 |
NM_144970.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009440.hg.1 |
NM_144970.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00009440.hg.1 |
NM_144970.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00164558.hg.1 |
ENST00000327877.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00164558.hg.1 |
ENST00000378421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00164558.hg.1 |
ENST00000378426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00164558.hg.1 |
NM_144970.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00164558.hg.1 |
ENST00000327877.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00164558.hg.1 |
ENST00000378421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00164558.hg.1 |
ENST00000378426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00164558.hg.1 |
NM_144970.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00164558.hg.1 |
TC0X00009440.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00009440.hg.1 |
ENST00000327877.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009440.hg.1 |
ENST00000327877.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009440.hg.1 |
ENST00000327877.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009440.hg.1 |
ENST00000327877.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009440.hg.1 |
ENST00000327877.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009440.hg.1 |
ENST00000327877.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009440.hg.1 |
ENST00000327877.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009440.hg.1 |
ENST00000327877.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009440.hg.1 |
ENST00000327877.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009440.hg.1 |
ENST00000378421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009440.hg.1 |
ENST00000378421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009440.hg.1 |
ENST00000378421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009440.hg.1 |
ENST00000378421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009440.hg.1 |
ENST00000378421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009440.hg.1 |
ENST00000378421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009440.hg.1 |
ENST00000378421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009440.hg.1 |
ENST00000378421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009440.hg.1 |
ENST00000378421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009440.hg.1 |
ENST00000378426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009440.hg.1 |
ENST00000378426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009440.hg.1 |
ENST00000378426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009440.hg.1 |
ENST00000378426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009440.hg.1 |
ENST00000378426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009440.hg.1 |
ENST00000378426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009440.hg.1 |
ENST00000378426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009440.hg.1 |
ENST00000378426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009440.hg.1 |
ENST00000378426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009440.hg.1 |
ENST00000378426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009440.hg.1 |
ENST00000378426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009440.hg.1 |
ENST00000378426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009440.hg.1 |
ENST00000378426.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00009440.hg.1 |
NM_144970.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009440.hg.1 |
NM_144970.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009440.hg.1 |
NM_144970.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009440.hg.1 |
NM_144970.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009440.hg.1 |
NM_144970.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009440.hg.1 |
NM_144970.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009440.hg.1 |
NM_144970.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009440.hg.1 |
NM_144970.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009440.hg.1 |
NM_144970.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00009440.hg.1 |
NM_144970.2 |
Refseq |