Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001063.hg.1 |
TC0X000081.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001063.hg.1 |
ENST00000380122.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001063.hg.1 |
ENST00000398155.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001063.hg.1 |
NM_001168683.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X001063.hg.1 |
NM_018360.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
ENST00000380122.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
ENST00000380122.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
ENST00000380122.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
ENST00000380122.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
ENST00000380122.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
ENST00000380122.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
ENST00000380122.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
ENST00000380122.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
ENST00000380122.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
ENST00000380122.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
ENST00000380122.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
ENST00000380122.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
ENST00000380122.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
ENST00000380122.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
ENST00000398155.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
ENST00000398155.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
ENST00000398155.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
ENST00000398155.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
ENST00000398155.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
ENST00000398155.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
ENST00000398155.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
ENST00000398155.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
ENST00000398155.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
ENST00000398155.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
ENST00000398155.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
ENST00000398155.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
ENST00000485153.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
ENST00000485153.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
ENST00000485153.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
ENST00000485153.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
ENST00000485153.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
ENST00000485153.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
NM_001168683.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
NM_001168683.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
NM_001168683.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
NM_001168683.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
NM_001168683.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
NM_001168683.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
NM_001168683.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
NM_001168683.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
NM_001168683.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
NM_001168683.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
NM_001168683.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
NM_001168683.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
NM_018360.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
NM_018360.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
NM_018360.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
NM_018360.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
NM_018360.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
NM_018360.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
NM_018360.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
NM_018360.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
NM_018360.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
NM_018360.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
NM_018360.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
NM_018360.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
NM_018360.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006701.hg.1 |
NM_018360.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
ENST00000380122.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
ENST00000380122.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
ENST00000380122.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
ENST00000380122.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
ENST00000380122.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
ENST00000380122.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
ENST00000380122.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
ENST00000380122.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
ENST00000380122.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
ENST00000380122.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
ENST00000380122.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
ENST00000380122.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
ENST00000380122.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
ENST00000380122.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
ENST00000398155.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
ENST00000398155.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
ENST00000398155.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
ENST00000398155.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
ENST00000398155.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
ENST00000398155.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
ENST00000398155.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
ENST00000398155.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
ENST00000398155.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
ENST00000398155.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
ENST00000398155.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
ENST00000398155.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
ENST00000485153.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
ENST00000485153.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
ENST00000485153.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
ENST00000485153.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
ENST00000485153.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
ENST00000485153.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
NM_001168683.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
NM_001168683.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
NM_001168683.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
NM_001168683.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
NM_001168683.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
NM_001168683.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
NM_001168683.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
NM_001168683.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
NM_001168683.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
NM_001168683.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
NM_001168683.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
NM_001168683.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
NM_018360.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
NM_018360.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
NM_018360.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
NM_018360.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
NM_018360.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
NM_018360.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
NM_018360.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
NM_018360.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
NM_018360.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
NM_018360.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
NM_018360.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
NM_018360.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
NM_018360.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006701.hg.1 |
NM_018360.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145896.hg.1 |
ENST00000380122.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145896.hg.1 |
ENST00000398155.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145896.hg.1 |
NM_001168683.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145896.hg.1 |
NM_018360.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145896.hg.1 |
ENST00000380122.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145896.hg.1 |
ENST00000398155.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145896.hg.1 |
NM_001168683.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145896.hg.1 |
NM_018360.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145896.hg.1 |
TC0X00006701.hg.1 |
Clariom D TC |