Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021095.hg.1 |
TC04001487.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021095.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021095.hg.1 |
ENST00000541197.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021095.hg.1 |
ENST00000545965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021095.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021098.hg.1 |
TC04001487.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021098.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021098.hg.1 |
ENST00000541197.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021098.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021102.hg.1 |
TC04001487.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021102.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021102.hg.1 |
ENST00000503013.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021102.hg.1 |
ENST00000541197.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021102.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021105.hg.1 |
TC04001487.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021105.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021105.hg.1 |
ENST00000503013.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021105.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021103.hg.1 |
TC04001487.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021103.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021103.hg.1 |
ENST00000503013.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021103.hg.1 |
ENST00000541197.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021103.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021104.hg.1 |
TC04001487.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021104.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021104.hg.1 |
ENST00000503013.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021104.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011650.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011650.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011650.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011650.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011650.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011650.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011650.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011650.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011650.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011650.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011650.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011650.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011650.hg.1 |
ENST00000503013.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011650.hg.1 |
ENST00000503013.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011650.hg.1 |
ENST00000503013.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011650.hg.1 |
ENST00000503013.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011650.hg.1 |
ENST00000503013.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011650.hg.1 |
ENST00000503013.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011650.hg.1 |
ENST00000503013.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011650.hg.1 |
ENST00000503013.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011650.hg.1 |
ENST00000509829.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011650.hg.1 |
ENST00000509829.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011650.hg.1 |
ENST00000509829.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011650.hg.1 |
ENST00000509829.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011650.hg.1 |
ENST00000509829.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011650.hg.1 |
ENST00000509829.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011650.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011650.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011650.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011650.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011650.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011650.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011650.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011650.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011650.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011650.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011650.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400011650.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021106.hg.1 |
TC04001487.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021106.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021106.hg.1 |
ENST00000503013.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021106.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021097.hg.1 |
TC04001487.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021097.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021097.hg.1 |
ENST00000541197.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021097.hg.1 |
ENST00000545965.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021097.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021107.hg.1 |
TC04001487.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021107.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021107.hg.1 |
ENST00000503013.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04021107.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007950.hg.1 |
ENST00000457003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007950.hg.1 |
ENST00000457003.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007950.hg.1 |
ENST00000457003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179996.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179996.hg.1 |
ENST00000503013.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179996.hg.1 |
ENST00000509829.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179996.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400179996.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179996.hg.1 |
ENST00000503013.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179996.hg.1 |
ENST00000509829.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179996.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400179996.hg.1 |
TC0400011650.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400179997.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179997.hg.1 |
ENST00000503013.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179997.hg.1 |
ENST00000509829.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179997.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400179997.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179997.hg.1 |
ENST00000503013.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179997.hg.1 |
ENST00000509829.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179997.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400179997.hg.1 |
TC0400011650.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400179999.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179999.hg.1 |
ENST00000503013.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179999.hg.1 |
ENST00000509829.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179999.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400179999.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179999.hg.1 |
ENST00000503013.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179999.hg.1 |
ENST00000509829.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179999.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400179999.hg.1 |
TC0400011650.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400011650.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011650.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011650.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011650.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011650.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011650.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011650.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011650.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011650.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011650.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011650.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011650.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011650.hg.1 |
ENST00000503013.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011650.hg.1 |
ENST00000503013.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011650.hg.1 |
ENST00000503013.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011650.hg.1 |
ENST00000503013.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011650.hg.1 |
ENST00000503013.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011650.hg.1 |
ENST00000503013.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011650.hg.1 |
ENST00000503013.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011650.hg.1 |
ENST00000503013.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011650.hg.1 |
ENST00000509829.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011650.hg.1 |
ENST00000509829.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011650.hg.1 |
ENST00000509829.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011650.hg.1 |
ENST00000509829.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011650.hg.1 |
ENST00000509829.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011650.hg.1 |
ENST00000509829.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400011650.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011650.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011650.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011650.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011650.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011650.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011650.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011650.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011650.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011650.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011650.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400011650.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400179995.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179995.hg.1 |
ENST00000503013.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179995.hg.1 |
ENST00000509829.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179995.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400179995.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179995.hg.1 |
ENST00000503013.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179995.hg.1 |
ENST00000509829.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179995.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400179995.hg.1 |
TC0400011650.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400180000.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180000.hg.1 |
ENST00000503013.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180000.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180000.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180000.hg.1 |
ENST00000503013.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400180000.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400180000.hg.1 |
TC0400011650.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400179991.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179991.hg.1 |
ENST00000509829.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179991.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400179991.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179991.hg.1 |
ENST00000509829.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179991.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400179991.hg.1 |
TC0400011650.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900007950.hg.1 |
ENST00000457003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007950.hg.1 |
ENST00000457003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007950.hg.1 |
ENST00000457003.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179998.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179998.hg.1 |
ENST00000503013.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179998.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400179998.hg.1 |
ENST00000315366.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179998.hg.1 |
ENST00000503013.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400179998.hg.1 |
NM_024590.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400179998.hg.1 |
TC0400011650.hg.1 |
Clariom D TC |