Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006308.hg.1 |
ENST00000222275.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006308.hg.1 |
ENST00000379013.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006308.hg.1 |
NM_007000.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19006308.hg.1 |
TC19000480.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
ENST00000222275.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
ENST00000222275.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
ENST00000222275.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
ENST00000222275.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
ENST00000222275.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
ENST00000222275.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
ENST00000222275.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
ENST00000222275.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
ENST00000222275.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
ENST00000379013.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
ENST00000379013.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
ENST00000379013.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
ENST00000379013.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
ENST00000379013.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
ENST00000379013.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
ENST00000379013.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
ENST00000379013.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
ENST00000379013.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
ENST00000616789.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
ENST00000616789.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
ENST00000616789.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
ENST00000616789.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
ENST00000616789.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
ENST00000616789.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
ENST00000616789.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
ENST00000616789.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
ENST00000616789.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
ENST00000616789.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
ENST00000617999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
ENST00000617999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
ENST00000617999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
ENST00000617999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
ENST00000617999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
ENST00000617999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
ENST00000617999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
ENST00000617999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
ENST00000617999.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
NM_001281443.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
NM_001281443.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
NM_001281443.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
NM_001281443.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
NM_001281443.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
NM_001281443.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
NM_001281443.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
NM_001281443.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
NM_001281443.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
NM_001281443.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
NM_007000.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
NM_007000.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
NM_007000.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
NM_007000.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
NM_007000.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
NM_007000.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
NM_007000.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
NM_007000.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007874.hg.1 |
NM_007000.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900158439.hg.1 |
ENST00000222275.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158439.hg.1 |
ENST00000379013.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158439.hg.1 |
ENST00000616789.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158439.hg.1 |
ENST00000617999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158439.hg.1 |
NM_001281443.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900158439.hg.1 |
NM_007000.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900158439.hg.1 |
TC1900007874.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900158439.hg.1 |
ENST00000222275.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158439.hg.1 |
ENST00000379013.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158439.hg.1 |
ENST00000616789.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158439.hg.1 |
ENST00000617999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900158439.hg.1 |
NM_001281443.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900158439.hg.1 |
NM_007000.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
ENST00000222275.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
ENST00000222275.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
ENST00000222275.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
ENST00000222275.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
ENST00000222275.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
ENST00000222275.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
ENST00000222275.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
ENST00000222275.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
ENST00000222275.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
ENST00000379013.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
ENST00000379013.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
ENST00000379013.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
ENST00000379013.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
ENST00000379013.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
ENST00000379013.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
ENST00000379013.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
ENST00000379013.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
ENST00000379013.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
ENST00000616789.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
ENST00000616789.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
ENST00000616789.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
ENST00000616789.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
ENST00000616789.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
ENST00000616789.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
ENST00000616789.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
ENST00000616789.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
ENST00000616789.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
ENST00000616789.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
ENST00000617999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
ENST00000617999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
ENST00000617999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
ENST00000617999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
ENST00000617999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
ENST00000617999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
ENST00000617999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
ENST00000617999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
ENST00000617999.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
NM_001281443.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
NM_001281443.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
NM_001281443.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
NM_001281443.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
NM_001281443.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
NM_001281443.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
NM_001281443.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
NM_001281443.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
NM_001281443.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
NM_001281443.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
NM_007000.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
NM_007000.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
NM_007000.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
NM_007000.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
NM_007000.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
NM_007000.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
NM_007000.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
NM_007000.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007874.hg.1 |
NM_007000.3 |
Refseq |