Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000316470.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000316470.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000316470.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000316470.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000316470.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000316470.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000316470.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000316470.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000316470.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000316470.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000316470.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000316470.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000316470.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000316470.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000416071.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000416071.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000416071.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000416071.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000416071.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000416071.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000420647.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000420647.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000420647.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000420647.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000420647.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000420647.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000420647.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000420647.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000420647.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000420647.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000420647.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000448950.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000448950.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000448950.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000448950.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000448950.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000448950.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000448950.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000448950.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000448950.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000448950.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000448950.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000448950.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000448950.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000489120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000489120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000489120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000489120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000489120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000489120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000489120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000489120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000489120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000489120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000489120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000489120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000489120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000489120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000489120.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000496739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000496739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000496739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000496739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000496739.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000621324.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000621324.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000621324.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000621324.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000621324.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000621324.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000621324.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000621324.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000621324.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000621324.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000622011.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000622011.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
ENST00000622011.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
NM_032604.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
NM_032604.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
NM_032604.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
NM_032604.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
NM_032604.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
NM_032604.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
NM_032604.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
NM_032604.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
NM_032604.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
NM_032604.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
NM_032604.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
NM_032604.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
NM_032604.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
NM_032604.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007054.hg.1 |
NM_032604.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002072.hg.1 |
TC02000165.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002072.hg.1 |
ENST00000316470.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002072.hg.1 |
NM_032604.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200148195.hg.1 |
ENST00000316470.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148195.hg.1 |
ENST00000416071.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148195.hg.1 |
ENST00000420647.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148195.hg.1 |
ENST00000448950.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148195.hg.1 |
ENST00000489120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148195.hg.1 |
ENST00000621324.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148195.hg.1 |
NM_032604.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200148195.hg.1 |
ENST00000316470.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148195.hg.1 |
ENST00000416071.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148195.hg.1 |
ENST00000420647.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148195.hg.1 |
ENST00000448950.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148195.hg.1 |
ENST00000489120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148195.hg.1 |
ENST00000621324.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200148195.hg.1 |
NM_032604.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200148195.hg.1 |
TC0200007054.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000316470.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000316470.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000316470.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000316470.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000316470.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000316470.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000316470.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000316470.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000316470.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000316470.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000316470.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000316470.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000316470.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000316470.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000416071.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000416071.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000416071.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000416071.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000416071.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000416071.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000420647.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000420647.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000420647.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000420647.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000420647.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000420647.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000420647.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000420647.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000420647.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000420647.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000420647.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000448950.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000448950.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000448950.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000448950.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000448950.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000448950.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000448950.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000448950.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000448950.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000448950.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000448950.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000448950.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000448950.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000489120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000489120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000489120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000489120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000489120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000489120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000489120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000489120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000489120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000489120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000489120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000489120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000489120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000489120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000489120.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000496739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000496739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000496739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000496739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000496739.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000621324.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000621324.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000621324.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000621324.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000621324.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000621324.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000621324.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000621324.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000621324.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000621324.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000622011.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000622011.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
ENST00000622011.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
NM_032604.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
NM_032604.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
NM_032604.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
NM_032604.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
NM_032604.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
NM_032604.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
NM_032604.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
NM_032604.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
NM_032604.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
NM_032604.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
NM_032604.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
NM_032604.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
NM_032604.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
NM_032604.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007054.hg.1 |
NM_032604.3 |
Refseq |