Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
ENST00000272771.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
ENST00000272771.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
ENST00000272771.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
ENST00000272771.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
ENST00000272771.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
ENST00000272771.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
ENST00000272771.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
ENST00000272771.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
ENST00000272771.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
ENST00000272771.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
ENST00000272771.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
ENST00000272771.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
ENST00000272771.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
ENST00000272771.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
ENST00000272771.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
ENST00000392314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
ENST00000392314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
ENST00000392314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
ENST00000392314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
ENST00000392314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
ENST00000392314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
ENST00000392314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
ENST00000392314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
ENST00000392314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
ENST00000392314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
ENST00000392314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
ENST00000392314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
ENST00000392314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
ENST00000392314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
ENST00000392314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
ENST00000392314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
ENST00000409056.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
ENST00000409056.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
ENST00000409056.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
ENST00000409056.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
ENST00000409056.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
ENST00000487771.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
ENST00000487771.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
ENST00000487771.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
NM_001305134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
NM_001305134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
NM_001305134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
NM_001305134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
NM_001305134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
NM_001305134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
NM_001305134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
NM_001305134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
NM_001305134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
NM_001305134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
NM_001305134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
NM_001305134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
NM_001305134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
NM_001305134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
NM_001305134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
NM_001305134.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
NM_001305145.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
NM_001305145.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
NM_001305145.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
NM_001305145.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
NM_001305145.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
NM_001305145.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
NM_001305145.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
NM_016192.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
NM_016192.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
NM_016192.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
NM_016192.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
NM_016192.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
NM_016192.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
NM_016192.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
NM_016192.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
NM_016192.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
NM_016192.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
NM_016192.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
NM_016192.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
NM_016192.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
NM_016192.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015266.hg.1 |
NM_016192.3 |
Refseq |