Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001447.2 | NP_001438.1 | 21 - 22 | 12068 | 77 | |
XM_006714761.3 | XP_006714824.1 | 22 - 23 | 12639 | 77 | ||
XM_011537600.2 | XP_011535902.1 | 24 - 25 | 12407 | 77 | ||
XM_011537603.2 | XP_011535905.1 | 22 - 23 | 12340 | 77 | ||
XM_017009224.1 | XP_016864713.1 | 24 - 25 | 12409 | 77 | ||
XM_017009225.1 | XP_016864714.1 | 23 - 24 | 12251 | 77 | ||
GenBank mRNA | AB011535.1 | - | 3 - 4 | 727 | 77 | |
AB160984.1 | - | 9 - 10 | 2378 | 77 | ||
AB177851.1 | - | 3 - 4 | 727 | 77 | ||
AF231022.1 | - | 21 - 22 | 12068 | 77 | ||
BC039143.2 | - | 4 - 5 | 1230 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |