Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033967.hg.1 |
TC02002101.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033967.hg.1 |
ENST00000342380.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033967.hg.1 |
NM_001002036.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033966.hg.1 |
TC02002101.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033966.hg.1 |
ENST00000342380.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033966.hg.1 |
NM_001002036.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033969.hg.1 |
TC02002101.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033969.hg.1 |
ENST00000342380.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02033969.hg.1 |
NM_001002036.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013510.hg.1 |
ENST00000342380.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013510.hg.1 |
ENST00000342380.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013510.hg.1 |
ENST00000342380.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013510.hg.1 |
ENST00000342380.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013510.hg.1 |
ENST00000342380.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013510.hg.1 |
ENST00000342380.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013510.hg.1 |
ENST00000342380.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013510.hg.1 |
ENST00000342380.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013510.hg.1 |
ENST00000342380.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013510.hg.1 |
ENST00000470582.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013510.hg.1 |
ENST00000470582.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013510.hg.1 |
ENST00000470582.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013510.hg.1 |
ENST00000470582.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013510.hg.1 |
ENST00000470582.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013510.hg.1 |
NM_001002036.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013510.hg.1 |
NM_001002036.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013510.hg.1 |
NM_001002036.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013510.hg.1 |
NM_001002036.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013510.hg.1 |
NM_001002036.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013510.hg.1 |
NM_001002036.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013510.hg.1 |
NM_001002036.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013510.hg.1 |
NM_001002036.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013510.hg.1 |
NM_001002036.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200198783.hg.1 |
ENST00000342380.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200198783.hg.1 |
ENST00000470582.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200198783.hg.1 |
NM_001002036.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200198783.hg.1 |
ENST00000342380.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200198783.hg.1 |
ENST00000470582.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200198783.hg.1 |
NM_001002036.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200198783.hg.1 |
TC0200013510.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200013510.hg.1 |
ENST00000342380.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013510.hg.1 |
ENST00000342380.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013510.hg.1 |
ENST00000342380.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013510.hg.1 |
ENST00000342380.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013510.hg.1 |
ENST00000342380.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013510.hg.1 |
ENST00000342380.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013510.hg.1 |
ENST00000342380.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013510.hg.1 |
ENST00000342380.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013510.hg.1 |
ENST00000342380.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013510.hg.1 |
ENST00000470582.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013510.hg.1 |
ENST00000470582.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013510.hg.1 |
ENST00000470582.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013510.hg.1 |
ENST00000470582.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013510.hg.1 |
ENST00000470582.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013510.hg.1 |
NM_001002036.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013510.hg.1 |
NM_001002036.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013510.hg.1 |
NM_001002036.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013510.hg.1 |
NM_001002036.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013510.hg.1 |
NM_001002036.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013510.hg.1 |
NM_001002036.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013510.hg.1 |
NM_001002036.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013510.hg.1 |
NM_001002036.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013510.hg.1 |
NM_001002036.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200198784.hg.1 |
ENST00000342380.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200198784.hg.1 |
ENST00000470582.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200198784.hg.1 |
NM_001002036.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200198784.hg.1 |
ENST00000342380.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200198784.hg.1 |
ENST00000470582.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200198784.hg.1 |
NM_001002036.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200198784.hg.1 |
TC0200013510.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200198786.hg.1 |
ENST00000342380.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200198786.hg.1 |
NM_001002036.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200198786.hg.1 |
ENST00000342380.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200198786.hg.1 |
NM_001002036.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200198786.hg.1 |
TC0200013510.hg.1 |
Clariom D TC |