Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000299194.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000299194.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000299194.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000299194.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000299194.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000299194.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000299194.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000299194.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000299194.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000299194.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000299194.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000299194.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000299194.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000299194.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000299194.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000299194.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000535041.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000535041.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000535041.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000535041.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000535041.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000535041.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000535041.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000535041.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000535041.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000535041.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000535041.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000535041.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000535041.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000539859.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000539859.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000539859.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000539859.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000539859.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000539859.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000540378.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000540378.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000540378.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000540378.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000540378.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000540378.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000542529.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000542529.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000542529.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000542529.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000542529.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000542529.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000543694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000543694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000543694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000543694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000543694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000543694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000543694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000543694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000543730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000543730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000543730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000543730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000543730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000543730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000543730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000543818.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000543818.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000543818.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000543818.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000543818.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000543818.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000543818.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000543818.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000543818.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000543818.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000543818.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000543818.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000543818.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000543818.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000543818.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000543818.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000543818.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000543818.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000543818.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000544489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000544489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000544489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000544489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000544489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000544489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000544489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000544489.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000545157.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000545157.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000545157.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000545157.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000545157.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000545157.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000545157.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000545157.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000546157.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000546157.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000546157.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000546157.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000546157.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000546157.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000546157.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000546157.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000546167.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000546167.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000546167.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000546167.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000546167.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000546167.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000546167.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
ENST00000546167.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001039029.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001039029.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001039029.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001039029.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001039029.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001039029.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001039029.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001039029.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001039029.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001039029.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001039029.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001039029.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001039029.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001039029.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001039029.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001039029.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001039029.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001039029.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001163925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001163925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001163925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001163925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001163925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001163925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001163925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001163925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001163925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001163925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001163925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001163925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001163925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001163925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001163925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001163925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001163925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001163925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001163925.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001163926.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001163926.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001163926.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001163926.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001163926.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001163926.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001163926.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001163926.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001163926.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001163926.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001163926.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001163926.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001163926.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001163926.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001163926.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001163926.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001163926.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006475.hg.1 |
NM_001163926.1 |
Refseq |